Learn More
Kimberly-Clark Professional™ Kimtech™ A5 Sterile Cleanroom Coveralls Product Code.: 11348041
Estimated Shipment: 14-07-2025
to see stock.
Add to basket
Kimberly-Clark Professional™ Kimtech™ A5 Sterile Cleanroom Coveralls
GREENER_CHOICE
Product Code. 11348041
4X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Product Code. 11348041
Brand: Kimberly-Clark Professional™ 88806
Estimated Shipment: 14-07-2025
to see stock.
Add to basket
Add to basket
Sterile cleanroom coveralls featuring innovative Clean-Don™ technology for simpler aseptic donning. Kimtech™ A5 Sterile Cleanroom Coveralls suitable for EU GMP ISO 5 Grade A sterile cleanrooms, the coveralls are individually vacuum-packed and made from light and breathable fabric.
Assured Compliance:
  • PPE Cat III according to Regulation (EU) 2016/425 (EU)
  • Type 5 and 6 certified
  • Certificate of conformance and sterility by lot available online
  • Suitable for use in EU GMP ISO 5 Class A sterile cleanroom environments and laboratories
  • Featuring advanced Clean-Don™ technology for fast, simple and compliant aseptic donning
  • Made from low-lint breathable SMS fabric with a cloth-like feel
  • Contamination protection is ensured through high quality sterility assured material with secure ties
  • Individually vacuum-packed and efficiently double-bagged to save space
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Product Type Cleanroom Coveralls
Size 4X-Large
Material SMS
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Snap
Seam Type Stitched
Sterility Sterile
Resistance None
Certifications/Compliance ISO 9001
Certifications/Compliance (Europe) EN 13034:2005 - Limited Splash Protection, EN ISO 13982-1:2004 - Particle Protection, CE CAT III, Category III PPE - CE Complex
Latex Free Yes
No. of Pockets None
Show More Show Less
  • KIMTECH - Reusable Apparel
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      KIMTECH - Reusable Apparel
    Product Content Correction
    Product Title
    Kimberly-Clark Professional™ Kimtech™ A5 Sterile Cleanroom Coveralls >