missing translation for 'onlineSavingsMsg'
Learn More
Kimberly-Clark Professional™ Kimtech™ A8 Clean Overalls Product Code.: 15558881
Add to basket

Kimberly-Clark Professional™ Kimtech™ A8 Clean Overalls
GREENER_CHOICE

Product Code. 15558881
Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy

Product Code. 15558881

Brand: Kimberly-Clark Professional™ 47693

Add to basket
Add to basket
This item is not returnable. View return policy

Maintain ISO 7 Class C or higher cleanroom environments with breathable, ultra light overalls. Kimtech™ A8 Clean Overalls are made of a high performance, chemical resistant fabric and come in compressed packaging to save space in the gowning room.

  • High performance chemical resistant fabric
  • Innovative, ultra light fabric: 45 gsm
  • Breathable
  • Highly comfortable
  • Compressed packaging to save space in the gowning room
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable

Specifications

Product Type Cleanroom Coveralls
Size Large
Material SMS
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Collared
Hood Type Unhooded
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance None
Collar Type Standing
Certifications/Compliance PPE Cat III Type 5/6 (Protection against particles/limited splash protection), EN1149-1:1995 Antistatic garment
For Use With (Application) Controlled Environments
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark Professional™ Kimtech™ A8 Clean Overalls >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.