Learn More
Kimberly-Clark Professional™ Kimtech™ M3 Pleat-Style Facemasks with Ties
Pleated style face mask with ties providing excellent bacterial and particle filtration to avoid contamination. Low-lint polyethylene film outer layer. Latex and silicone free. Suitable for use in ISO 3 or higher cleanrooms.
Brand: Kimberly-Clark Professional™ 62466
163.72 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Excellent bacterial and particle filtration to avoid contamination.
- Low-lint polyethylene film outer layer.
- Latex and silicone free.
- Suitable for use in ISO 3 or higher cleanrooms.
Specifications
Cleanroom Face Mask | |
Polyethylene | |
ISO 14001, ISO 9001; CE CAT I; Industrial Use Only | |
Yes |
White | |
Non-sterile | |
Soft Ties | |
62466 |