missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Kinase buffer I (5X)
For cellular and molecular biology applications
£44.12 - £53.16
Specifications
Components | 100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4. |
---|---|
Content And Storage | Ambient temperatures |
Product Type | Kinase buffer I (5X) |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15444449
|
Thermo Scientific™
J61967.AK |
250 mL |
£53.16
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15434449
|
Thermo Scientific™
J61967.AE |
100 mL |
£44.12
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4
Specifications
100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4. | |
Ambient temperatures | |
Kinase buffer I (5X) | |
Liquid |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title