Learn More
Kinase Buffer T (5X)
Kinase Buffer T (5X)
Brand:  Invitrogen PV5373
Code : 50
Additional Details : Weight : 0.50000kg
Product Code. 10092803
  • £118.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
Kinase buffer that has been specially optimized for use with the Type III PI4 kinases including PI4KA and PI4KB.
Specifications
Specifications
5X
1 Each
Liquid
Pharma and biopharma, target and lead identification and validation
4 mL