missing translation for 'onlineSavingsMsg'
Learn More

Kinase Quench Buffer

Kinase Quench Buffer

Brand:  Invitrogen P2825

Product Code. 10033343

  • £74.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Kinase Quench Buffer (500 mM EDTA) is an essential component for kinase assays using LanthaScreen™ technology. The Kinase Quench Buffer is used to stop the kinase reaction.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2213680C Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberP2825
Lot Number2213680B Certificate TypeCertificate of Analysis Date06/02/2023 Catalog NumberP2825
Lot Number1885967B Certificate TypeCertificate of Analysis Date12/02/2020 Catalog NumberP2825
Lot Number1966083C Certificate TypeCertificate of Analysis Date24/10/2018 Catalog NumberP2825
Lot Number1966083B Certificate TypeCertificate of Analysis Date23/02/2018 Catalog NumberP2825
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kinase Quench Buffer > 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.