missing translation for 'onlineSavingsMsg'
Learn More

HMC Europe Vertical Autoclave Baskets

With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.

Brand:  HMC Europe 50008

 View more versions of this product

Product Code. 10769754

  • £389.00 / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For sue with the HMC-HG and HMC-HV autoclaves

Specifications

Specifications

15.35 in.
22.45 gal.
HV 85 autoclave
29 cm
Stainless Steel
39 cm
85 L
11.41 in.
Stackable basket
390 x 290 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.