missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMC Europe Vertical Autoclaves: HV Series
Programmable top-loading autoclaves with varying chamber volumes
£14170.98 - £17065.86
Specifications
Certifications/Compliance | CE |
---|---|
For Use With (Application) | Steam sterilization |
Product Code | Brand | Capacity (Metric) | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Model | Price | Quantity & Availability | ||||
10448922
|
HMC Europe
20003 |
110 L | HV L110 |
£17065.86
Each |
Estimated Shipment: 28-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10693211
|
HMC Europe
20002 |
85 L | HV L85 |
£14170.98
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
Description
- Use steam sterilization
- Six sizes from 25 to 110 liters
- Low height, easy-to-load
- Portable — require no plumbing to operate but can be plumbed
- Complete line of baskets, inserts and cans available separately
- HV Series (dependable, hard-working, economical)
- Many upgrades available depending on your needs
Certifications
CE marked
Warranty
Two years on parts; One year on labor
Specifications
CE | |
Steam sterilization |