Learn More
Kinematica Polytron™ Homogenizer Generator Tool
Shop All Kinematica Products
Click to view available options
For Use With (Equipment):
Polytron™ generators
Unit Size:
Each
Description
- Fits all Polytron™ generators between 5 and 60 mm
- Precision made by skilled Swiss craftsmen
Specifications
Specifications
| Description | Homogenizer tool |
| For Use With (Application) | Changing rotors on homogenizers |
| For Use With (Equipment) | Polytron™ generators |
Product Content Correction
Product Title
Kinematica Polytron™ Homogenizer Generator Tool >
Spot an opportunity for improvement?Share a Content Correction