Learn More
Macherey-Nagel™ KingFisher™ 24 Accessory Kit
Enables automated purification of genomic DNA using a NucleoMag™ kit on the KingFisher™ Flex platform for faster, high throughput performance with reliable yields.
Brand: Macherey-Nagel™ 744953
Additional Details : Weight : 0.30000kg
Description
- Adapted NucleoMag™ procedure for instruments of the KingFisher™ Flex series
- Reliable performance and excellent yields from different sample materials
- Speeds up nucleic acid purification workflow: process 24 samples at once
- Set includes Deep-well Plates and Tip Combs for processing in 24-well format
Specifications
KingFisher™ Flex platform, NucleoMag™ kit | |
Automated nucleic acid purification | |
Deep-well Plates, Tip Combs for processing in 24-well format | |
1 Set |