Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Brand:  Thermo Scientific™ 95040460
227.55 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
 View more versions of this product
Product Code. 10329827
  • £249.00 / Pack of 50
Explore more special offers
Description
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 Deep-well Plate
96 Samples
KingFisher 96 deep-well plate, sterile (for Duo Prime, Flex and Presto)
KingFisher Duo Prime, Flex, and Presto
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.