Learn More
Thermo Scientific™ KingFisher Duo Prime Accessories
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources.
£259.00 - £458.00
Specifications
Product Type | Nucleic Acid Extraction and Cell Separation Instrument |
---|---|
Sample Type | Fresh or Frozen Plant Tissues, FFPE Tissue, Blood, Cells, Plasma |
Disposable | Reusable |
For Use With (Equipment) | KingFisher Duo Prime Purification System |
Product Code | Brand | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Price | Quantity & Availability | |||||
30132313
|
Thermo Scientific™
N12460 |
24 Deep-well Head |
£458.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30131189
|
Thermo Scientific™
N12459 |
96 Deep-well Head |
£450.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30131520
|
Thermo Scientific™
N12462 |
24 Deep-well Heating Block |
£259.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30131332
|
Thermo Scientific™
N12461 |
96 Deep-well Heating Block |
£262.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality.
The KingFisher Duo Prime instrument comes with a 12-pin magnet head for 96 deep well applications, a 6-pin magnet head for 24 deep well applications, and heating blocks. Additional magnet heads and heating blocks are offered here for separate purchase.Specifications
Nucleic Acid Extraction and Cell Separation Instrument | |
Fresh or Frozen Plant Tissues, FFPE Tissue, Blood, Cells, Plasma | |
Reusable | |
KingFisher Duo Prime Purification System |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.