Learn More
Macherey-Nagel™ KingFisher™ Duo Prime Accessory Kit
Enables automated purification of genomic DNA using a NucleoMag™ kit on the KingFisher™ Duo / Duo Prime platform for faster, high throughput performance with reliable yields.
Brand: Macherey-Nagel™ 744952
Additional Details : Weight : 0.40000kg
Description
- Adapted NucleoMag™ procedure for instruments of the KingFisher™ Duo/Duo Prime series
- Reliable performance and excellent yields from different sample materials
- Speeds up nucleic acid purification workflow: process 96 samples in less than 30 minutes (excluding lysis)
- Set includes Deep-well Plates, Tip Combs and Elution Strips for processing in 96-well format
Specifications
KingFisher™ Duo/Duo Prime platform, NucleoMag™ kit | |
Automated nucleic acid purification | |
Deep-well Plates, Tip Combs and Elution Strips for processing in 96-well format | |
1 Set |