Learn More
Thermo Scientific™ KingFisher™ Duo-Ready DNA Ultra2.0 Prefilled Plates
Automation-ready plates prefilled with MagMAX DNA Multi-Sample Ultra 2.0 reagents suitably configured for KingFisher Duo and KingFisher Duo Prime purification systems.
Brand: Thermo Scientific™ A36584
627.26 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 1.21600kg
Description
- KingFisher Duo-Ready DNA Ultra 2.0 Prefilled Plates enable isolation of genomic DNA from 50 to 400 μL of multiple sample types such as whole blood, saliva, buffy coat, buccal swabs, and more
- The purified gDNA is ideal for a variety of downstream molecular biology applications
- Key features of the KingFisher Duo-Ready DNA Ultra 2.0 Prefilled Plates:
- Maximize convenience with MagMAX DNA Multi-Sample Ultra 2.0 isolation chemistry prefilled into KingFisher SBS microtiter plates
- Increase ease of use—process 12 samples on instrument in 45 minutes without the hassle of reagent setup
- Designed to reduce user error
- Helps to reduce hands-on time
- Formatted specifically for KingFisher Duo instruments
- All the benefits of MagMAX sample prep chemistries in automation-ready plate format
- Maximize convenience and confidence with ready-to-use prefilled plates and accessory plastics configured specifically for the KingFisher Duo Prime instrument
- Eliminate the need to aliquot magnetic beads, lysis solutions, and wash solutions. Just peel off the foil seal from the KingFisher Duo-Ready DNA Ultra 2.0 prefilled plate; add enhancer, 50-400 μL of your sample, and proteinase K; and load the plate into the KingFisher Duo Prime instrument
- Eliminate the need for pipetting reagents, searching for plastics, or risking reagent mix-ups in your DNA isolation workflow
- What you get:
- Eight foil-sealed KingFisher Duo-Ready DNA Ultra 2.0 Prefilled Plates sufficient to process 12 samples in each instrument run (95 samples in total)
- Eight complete sets of accessory plastics (KingFisher 12-tip comb, elution strips, and caps)
- Proteinase K solution, enhancer solution, and elution solution sufficient for 96 samples
Specifications
Room Temperature | |
KingFisher Duo and KingFisher Duo Prime purification systems | |
DNA extraction | |
96 Preps |