Learn More
Thermo Scientific™ KingFisher™ Flex 24 Deep Well head
The KingFisher™ Flex 24 Deep Well head a magnetic head for the KingFisherTM Flex Magnetic Particle Purification System (Cat. Nos. 5400630 and 5400640). This magnetic head can serve as a replacement head for the 24 deep-well format system.
Brand: Thermo Scientific™ 24074440
3610.00 GBP valid until 2024-06-28
Use promo code "21689" to get your promotional price.
Code : 50
Additional Details : Weight : 0.01000kg
Description
The KingFisher™ Flex 24 Deep Well head a magnetic head for the KingFisherTM Flex Magnetic Particle Purification System (Cat. Nos. 5400630 and 5400640). This magnetic head can serve as a replacement head for the 24 deep-well format system. Both deep-well plates and KingFisher 96 plates can be used during the same run. Therefore, it is possible to start the processing by using larger volumes (in a deep-well plate) and elute the purified sample to a smaller volume (in a KingFisher 96 or MagMAX 96 standard plate).
The KingFisher™ Flex 24 Deep Well head a magnetic head for the KingFisherTM Flex Magnetic Particle Purification System (Cat. Nos. 5400630 and 5400640). This magnetic head can serve as a replacement head for the 24 deep-well format system. Both deep-well plates and KingFisher 96 plates can be used during the same run. Therefore, it is possible to start the processing by using larger volumes (in a deep-well plate) and elute the purified sample to a smaller volume (in a KingFisher 96 or MagMAX 96 standard plate).Specifications
24 Deep-well Head | |
KingFisher™ Flex | |
Room Temperature |
24 Samples | |
KingFisher™ |
For Research Use Only. Not for use in diagnostic procedures.