Learn More
Thermo Scientific™ KingFisher™ Flex Deep Well heating block
The KingFisher™ Flex 96 Deep-Well Heating Block is compatible with the KingFisher™ Flex Microtiter Deep-Well 96 Plate and the MagMAX™ Express-96 Deep-Well Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head.
Brand: Thermo Scientific™ 24075430
Code : 50
Additional Details : Weight : 0.01000kg
Specifications
96 Deep-well Plate | |
KingFisher™ Flex | |
Room Temperature |
96 Samples | |
KingFisher™ |
For research use only. Not for use in diagnostic procedures.