Learn More
Thermo Scientific™ KingFisher™ Flex 96 heating block
The KingFisher™ Flex 96 KF Heating Block is compatible with the KingFisher™ 96 KF Microplate (200 μL) and the MagMAX™ Express-96 Standard Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head.
Brand: Thermo Scientific™ 24075420
451.25 GBP valid until 2024-06-28
Use promo code "21689" to get your promotional price.
Code : 50
Additional Details : Weight : 0.01000kg
Description
The KingFisher™ Flex 96 KF Heating Block is compatible with the KingFisher™ 96 KF Microplate (200 μL) and the MagMAX™ Express-96 Standard Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head. This heating block is also compatible with the KingFisher™ Flex Microtiter Deep Well 96 Plate and the MagMAX™ Express-96 Deep Well plates.Specifications
96-well Plate | |
KingFisher™ Flex | |
Room Temperature |
96 Samples | |
KingFisher™ |
For research use only. Not for use in diagnostic procedures.