missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex Purification System

Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source.

Brand:  Thermo Scientific™ 5400620

 View more versions of this product

Product Code. 11839520

  • POA / Each

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.

Find a flexible solution for different types of sample processing needs. With 24 configurations, customers can raise the processing volume up to 5mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.

• Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
• High throughput system processes up to 96 samples in less than 20 minutes.
• Expands sample volume by 10-fold with the 24-well format.
• Open and flexible system lets the customer use anymagnetic particle-based kit tomeet the application demands.
• Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation andmodification.
• Ready-made protocols for different type of applications are available.

Recommended for:
• Genomics and proteomics
• Target identification
• Veterinary assays
• Biomarker discovery
• Quality control
• Forensics

TRUSTED_SUSTAINABILITY
Specifications

Specifications

20 to 200 μL
96-well Plate
KingFisher 96 Plate
START / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / four cursor keys / LCD
From +5°C above ambient temperature to +115°C
ca. >1 μm
23.6 in.
15 in.
62 lb.
26.8 in.
24 or 96
96 Samples
> 95%
Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
96 in one frame; Interchangeable magnet heads
24 or 96-well plates: KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 plate (20 to 200 μL), PCR plate (20 to 100 μL)
60 cm
38 cm
28 kg
68 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number711-8GF00363 Certificate TypeCertificate of Compliance Date28/01/2025 Catalog Number5400620
Lot Number711-8GF00364 Certificate TypeCertificate of Compliance Date28/01/2025 Catalog Number5400620
Lot Number711-8GF00396 Certificate TypeCertificate of Compliance Date27/01/2025 Catalog Number5400620
Lot Number711-8GF00397 Certificate TypeCertificate of Compliance Date27/01/2025 Catalog Number5400620
Lot Number711-8GF00398 Certificate TypeCertificate of Compliance Date27/01/2025 Catalog Number5400620
Special Offers

Special Offers

  • Multiple Manufacturers
    Fall in love with big savings
    Promo Code: 24107

    Expires2025-03-31

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ KingFisher™ Flex Purification System > KingFisher Flex with 96 KF head

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use