Learn More
Thermo Scientific™ KingFisher™ Flex-Ready DNA Ultra 2.0 Prefilled Plates
Automation-ready plates prefilled with MagMAX DNA Multi-Sample Ultra 2.0 reagents suitably configured for KingFisher Flex purification systems
Brand: Thermo Scientific™ A36586
647.08 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 1.32400kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Enable isolation of genomic DNA from 50 to 400 μL of multiple sample types such as whole blood, saliva, buffy coat, buccal swabs, and more. The purified gDNA is ideal for a variety of downstream molecular biology applications
- Key features of the KingFisher Flex-Ready DNA Ultra 2.0 Prefilled Plates:
- Maximize convenience with MagMAX DNA Multi-Sample Ultra 2.0 isolation chemistry prefilled into KingFisher SBS microtiter plates
- Increase ease of use—process 96 samples on instrument without the hassle of reagent setup
- Designed to reduce user error
- Helps to reduce hands-on time
- Formatted specifically for KingFisher Flex instruments
- All the benefits of MagMAX sample prep chemistries in automation-ready plate format
- Eliminate the need to aliquot magnetic beads, lysis solutions, and wash solutions. Just peel off the foil seals from the KingFisher Flex-Ready DNA Ultra 2.0 prefilled plates; add enhancer, 50-400 μL of your sample, and proteinase K; and load all of the plates into the KingFisher Flex instrument
- Eliminate the need for pipetting reagents, searching for plastics, or risking reagent mix-ups in your DNA isolation workflow
- Complete, prefilled automation-ready sample prep kit improves convenience with all accessory plastics
- What you get:
- Seven foil-sealed KingFisher Flex-Ready DNA Ultra 2.0 Prefilled Plates sufficient to process 96 samples in one instrument run (96 samples in total)
- One empty and sealed sample input 96 deep well plate
- KingFisher 96-tip comb
- Proteinase K solution and enhancer solution sufficient for 96 samples
Specifications
Room Temperature | |
KingFisher Flex purification systems | |
DNA extraction | |
96 Preps |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- LONG-TERM AQUATIC HAZARD Chronic 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P270-Do not eat, drink or smoke when using this product.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.