Learn More
Thermo Scientific™ KingFisher™ Purification System
Automate time-consuming sample preparation processes of nucleic acids, proteins and cells with Thermo Scientific™ KingFisher™ Purification Systems.
Brand: Thermo Scientific™ 5400000
Description
The KingFisher™ Purification System is pending discontinuation. Thermo Fisher Scientific offers other automation platforms such as the KingFisher Duo Prime Purification System, KingFisher Flex Purification System and KingFisher Apex Purification System. The company will continue to support current KingFisher Purification System users with plastics, service, and technical support through 2028, or best effort.
By August 1st, 2023, Thermo Fisher Scientific will transition to limited support stage. It will offer limited support contracts and provide best-effort service and support, subject to availability of spare parts and resources.
If you are interested in upgrading to one of its newer sample preparation instruments, please contact your local account manager for product information.
Features:- Offer rapid and reproducible purification of high-quality DNA, RNA, proteins and cells for various types of downstream applications
- Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands
- Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification
- Ready-made protocols for different types of applications available
Recommended for:
Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays
Specifications
20 to 200 μL | |
Kingfisher Purification System, 100-240V, 50/60Hz | |
START/STOP/TWO CURSOR KEYS/LCD | |
2 x 12 format | |
Each | |
Special design, 8 x 12 grid format | |
29 cm | |
31 cm | |
10 kg | |
29 cm |
24 Samples/Run | |
99% | |
Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays | |
ca. >1 μm | |
Special design, 2 x 12 format | |
11.4 in. | |
12.2 in. | |
23 lb. | |
11.4 in. | |
100-240 V, 50/60 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For research use only. Not for use in diagnostic procedures.