Learn More
Thermo Scientific™ KingFisher™ Presto Purification System
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
Brand:  Thermo Scientific™ 24078841
 View more versions of this product
Product Code. 15861647
  • £4085.00 / Each
Explore more special offers
Description
Description
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories. Utilizing proven KingFisher magnetic particle-based technology, the KingFisher Presto provides high-quality yields of target nucleic acids and proteins for biopharma, biotech and research.
Features:
• Purify 96 or 24 samples at a time
• Volume range of 50–5000 μL
• Minimized size for easy fitting to automation systems
• Heating capability for application requiring elevated temperatures
• Easy connection to several liquid handling instruments
• Flexible system for wide variety of applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 to 5000 μL
KingFisher Presto 24 DW head with heating block
24