Learn More
Thermo Scientific™ KingFisher™ Systems Consumables
Optimize performance using Thermo Scientific™ KingFisher™Systems Consumables, ideal for magnetic particle processing.
Brand: Thermo Scientific™ 97002084
Code : 50
Additional Details : Weight : 3.92810kg
Description
Optimize performance using Thermo Scientific™ KingFisher™ Systems Consumables, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially-designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
- Specifically designed for use with the KingFisher System
- Ideal for magnetic particle processing because of their low binding affinity for biomolecules
- The specially-designed, disposable tip combs and microplates are made of polypropylene
- This enables excellent recovery of magnetic beads
Specifications
200 μL | |
Magnetic particle processing | |
50/Cs. |
KingFisher plate 200 μL, 50 pcs/box | |
KingFisher System |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.