Learn More
Kjeldahl Boric Acid Solution, Pure, Solution 4% Alkaline with Indicator, Fisher Chemical™
Brand: Fisher Chemical K/0200/21
170.49 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 23
Additional Details : CAS Number : 10043-35-3 Weight : 5.00000kg
Specifications
Red | |
5 L | |
Plastic Container |
Safety and Handling
- Kjeldahl 4% Boric acid solution with indicator
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Boric acid, Methyl red