missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical™
Click to view available options
Quantity:
1000 Tablets
500 Tablets
Description
Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical, Quantity: 1000 tablets, Blue-White, MDL Number: 147336, Packaging: Plastic powder jar, Physical Form: Solid
Specifications
Specifications
Color | Blue-White |
Packaging | Plastic powder jar |
Quantity | 500 Tablets |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'