missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl Reagent Boric Acid 2% w/v Solution without Indicator, Reagecon

Kjeldahl Reagent Boric Acid 2% w/v Solution without Indicator

Brand:  Reagecon BOA25

Product Code. 11583182

  • £108.00 / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Kjeldahl Reagent Boric Acid 2% w/v Solution without Indicator
Specifications

Specifications

5 L
HDPE Container
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers