missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl Sodium Hydroxide/Sodium Thiosulfate Solution, Technical, for Kjel-Foss Automatic Nitrogen Analyser, Fisher Chemical

Product Code. 10704381
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

MDL Number 60749
Quantity 5 L
Packaging Plastic container
Physical Form Liquid
compliance-icons
Product Identifier
  • Kjeldhal Sodium hydroxide/sodium thiosulfate solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'