Learn More
Kjeldahl tablets with selenium (Wieninger), Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 243920010
144.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Kjeldahl tablets with selenium (Wieninger) | |
1 kg | |
Plastic bottle |
7782-49-2, 7758-99-8, 7757-82-6 | |
MFCD00147338 |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Kjeldahl tablets with selenium (Wieninger)
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468058 | Certificate of Analysis | 31/01/2025 | 243920010,243920025 |
A0468059 | Certificate of Analysis | 31/01/2025 | 243920010,243920025 |
A0460383 | Certificate of Analysis | 19/02/2024 | 243920010,243920025 |
A0455918 | Certificate of Analysis | 25/08/2023 | 243920010,243920025 |
A0453058 | Certificate of Analysis | 25/08/2023 | 243920010,243920025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only