Learn More
KjTabs VCT - Kjeldahl Catalyst Tablets, Velp Scientifica™
KjTabsTM VCT Kjeldahl Catalyst Tablets are ideal for tougher samples. The perfect solutions to speed up the digestion process.
Brand: Velp Scientifica A00000276
This item is not returnable.
View return policy
Description
The right KjTabsTM to fit your needs
The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses. Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with VELP Scientifica genuine catalyst tablets.
The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
The KjTabs™ VCT consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Titanium (Ti) as metal salts to accelerate the mineralization process.
VCT for tougher samples & even faster digestion
5g K2SO4 + 0,15g CuSO4 x 5 H2O + 0,15g TiO2
1000 pcs
The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
The KjTabs™ VCT consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Titanium (Ti) as metal salts to accelerate the mineralization process.
VCT for tougher samples & even faster digestion
5g K2SO4 + 0,15g CuSO4 x 5 H2O + 0,15g TiO2
1000 pcs
VELP analytical experts KjTabs™ catalyst tablets application suggestions
The KjTabs™ VCT are a suitable choice when performing Kjeldahl analysis for Nitrogen determination in milk and dairy products.
Specifications
1000 Pc./Box |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Kjeldahl Catalyst Tablets
Signal Word
- Warning
Hazard Category
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Copper Sulphate Pentahydrate