missing translation for 'onlineSavingsMsg'
Learn More

KjTabs VKPC - Kjeldahl Catalyst Tablets, Velp Scientifica™

KjTabsTM VKPC Kjeldahl Catalyst Tablets are ideal when a high amount of copper is required. The perfect solutions to speed up the digestion of your samples.

Brand:  Velp Scientifica A00000275


Product Code. 15541775

  • £206.00 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The right KjTabs™ to fit your needs

 
The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses. Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with VELP Scientifica genuine catalyst tablets.

The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.

The KjTabs™ VKPC consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) as metal salt to accelerate the mineralization process.

VKPC ideal when high amount of copper is required
4,5g K2SO4 + 0,5g CuSO4 x 5 H2O
1000 pcs
 
 

VELP analytical experts KjTabs™ catalyst tablets application suggestions

The KjTabs™ VKPC are the most suitable choice when performing Kjeldahl analysis for Protein determination in bread and baked products.
Specifications

Specifications

1000 Pc./Box
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • KJTABS VKPC

Signal Word
  • Warning

Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : copper sulphate pentahydrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers