Learn More
KjTabs VW - Kjeldahl Catalyst Tablets, Velp Scientifica™
KjTabsTM VW Kjeldahl Catalyst Tablets are ideal for applications based on the Wieninger method. The perfect solutions to speed up the digestion of your samples.
Brand:  Velp Scientifica A00000282
Product Code. 15571775
  • £354.00 / Pack of 1000
Explore more special offers
Description
Description
The right KjTabs™ to fit your needs
The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses. Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with VELP Scientifica genuine catalyst tablets.
The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
The KjTabsTM VW consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Selenium (Se) as metal salts to accelerate the mineralization process.
VW recommended for applications based on the Wieninger method
4,875 g K2SO4 + 0,075 g CuSO4 x 5 H2O + 0,050 g Se
1000 pcs
Specifications
Specifications
1000 Pc./Box
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : copper sulphate pentahydrate, selenium