Learn More
SCHOTT AG Lighting and Imaging™ KL 300 LED Cold Light Source
Innovative fiber optic LED Light Source uses cold light to illuminate all types of objects in life sciences and industrial applications. The brightness equivalent to a 30-watt halogen lamp is ideal as a standard illumination system for the requirements of routine inspections and education.
Brand: SCHOTT AG Lighting and Imaging™ 120300
This item is not returnable.
View return policy
Description
- 80 Lm lightflux.
- No fan, silent operation.
- Compact ergonomical design.
- Mountable direct to the stand or pillar.
- Adaptors for all stands plus stand-alone version available.
- Mounting bracket need to be ordered separately.
- Wide range power supply included with international clip plug system.
- Fiber optic light guides or ringlights need to be ordered separately.
Specifications
Cold Light Source | |
107 x 114 x 61 mm | |
SCHOTT Fiber Optic Flexible Lightguides, Goosenecks or Ringlights with active lightguide diameter of max. 6 mm | |
2.4 in. | |
Wide range power supply with international clip plug system | |
4.2 in. | |
1 High Power LED white | |
100/240 V | |
114 mm |
CE Certified, B EMC-Class | |
Stereo Microscopy Illumination | |
50/60 Hz | |
61 mm | |
50,000 Hours | |
107 mm | |
80 Lm | |
4.5 in. |