missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 42-474 Bonded Nitrile-Coated Full-Length Heat Barriers

Non-woven felt insulation designed for intermittent handling of dry objects up to 200°C (400°F). Bonded nitrile coating protects against snags, punctures, abrasions, and cuts.

Brand:  Ansell 42474090

Code : NEW

Additional Details : Weight : 1.83600kg

 View more versions of this product

Product Code. 15692924

  • £241.00 / 12 pairs
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

  • Two-piece construction eliminates outseams for improved finger comfort, dexterity, and flexibility
  • Bonded nitrile coating protects against snags, punctures, abrasions, and cuts
  • Designed for intermittent handling of dry objects up to 200°C (400°F)
  • Enhanced grip capabilities when handling dry or oily surfaces
  • Compliant to FDA food handling requirements
  • Sweat absorbent non-woven felt inner
  • Asbestos-free
Specifications

Specifications

EN 420:2003 + A1:2009; Category III
Gray
Polyester
480°F
2241B
Gauntlet
ActivArmr™ 42-474
Nitrile
ANSI A2
9
250°C
Parts Handling, Press Injection Molding, Testing, and Welding
Felt
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers