Learn More
Kimberly-Clark™ Professional KleenGuard™ A10 Light Duty Overshoe With Sole
Made from SMS anti-static material with robust vinyl sole and elasticated upper for enhanced protection.
Brand: Kimberly-Clark™ Professional 82720
258.09 GBP valid until 2024-12-31
Use promo code "22916" to get your promotional price.
Code : 02
Additional Details : Weight : 0.50000kg
Description
- Single-use overshoe/boot constructed using the A50 Anti-static base sheet with robust sole
- Breathable, non-absorbent elasticated upper plus vinyl sole
- Improves safety and comfort in high-risk areas
- Protects against water-based splashes, liquids or dust particles and provides general protection
- Ideal for: keeping footwear clean and covered, helping to reduce contamination in food processing and light industrial areas, protecting safety footwear from splashes and spills in heavier industrial areas
Specifications
White | |
16.107 in. | |
12.694 lb. | |
A10 Light Duty Overshoe With Sole |
41.3 x 22.1 cm | |
41.3 cm | |
5.77 kg |