missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ KleenGuard™ A40 Liquid and Particle Protection Coveralls
173.19 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Color: White
Description
- Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
- Single use and breathable protective clothing designed for maximum comfort and safety, providing anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
- Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
- Silicone-free, low lint and treated to be antistatic - to EN 1149-5
- Ideal for protecting against powders, larger fibres and protection against a wide range of chemicals - reducing contamination in critical areas
- Storm flap and hood designed for easier taping to a respirator
- Suitable for use in pharmaceutical industries, manufacturing, utilities, electronics, agriculture and paint spraying areas
Specifications
Product Type | Hooded Coveralls |
Size | Small |
Material | KleenGuard™ |
Color | White |
Hood Type | Respirator Fit |
Protection Class | A40 |
For Use With (Application) | Providing high levels of protection against larger particles, radioactive dust and chemical splash |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'