missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ KleenGuard™ A50 Breathable Splash and Particle Protection Jacket

Product Code. 12621446
Large
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Single-use, breathable jacket with specially designed hood to fit a respirator. KleenGuard™ A50 Breathable Splash and Particle Protection Jacket is designed for comfort, safety and general protection against larger particles, radioactive dust and chemical splashes.

  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
  • Single use and breathable protective clothing designed for maximum comfort and safety, providing anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
  • Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
  • Silicone-free, ultra-low lint and treated to be antistatic - to EN 1149-5
  • Ideal for protecting against powders, larger fibres and protection against a wide range of chemicals reducing contamination in critical areas
  • Suitable for use in pharmaceutical industries, manufacturing, utilities, electronics, agriculture and paint spraying areas
  • Storm flap and hood designed for easier taping to a respirator

Specifications

Product Type Chemical Resistant Lab Coat
Size Large
Material Spunbond Polypropylene
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Puncture, Flame, Abrasion, Tears, Particulates, Liquids
Style Full-length
No. of Pockets None
For Use With (Application) Very good protection against particles, radioactive dust and chemical splashes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ KleenGuard™ A50 Breathable Splash and Particle Protection Jacket >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.