missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ A7 White Laboratory Coats

Kimtech™ A7 Laboratory Coats are breathable, multi-layered lab coats offering Type 6 limited chemical splash protection. The latex-free coats are chemical- and abrasion-resistant and have a comfortable cloth-like feel.

Specifications

Product Type General Purpose Lab Coat
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
View More Specs

Description

Description

  • Assured Compliance:
    • PPE Cat III according to (EU) Regulation 2016/425: Type 6 (PB) limited chemical protection
    • Abrasion Resistance Class 2 (visual) EN 530 M2
    • Tensile Strength Class 1 EN ISO 13934-1
    • Class 3 Resistance to Penetration by Liquid Chemicals; 10% NaOH Class 3, EN 368 30% H2SO4
    • Resistance to Penetration by Liquids (spray test) Not Tested (parts of the body) EN 468 (modified)
  • High quality disposable laboratory coats with triple stitched seams and breathable, multi-layered material.
  • Three fabric layers provide enhanced protection against chemical splash, dry particulates and a variety of water-based liquids.
  • Made from breathable and abrasion-resistant layers and featuring triple over-locked seams for added strength and durability.
  • Latex-free, silicon-free and low lint construction.
  • Snap fastening
  • Three fabric layers for enhanced protection.
  • Individually packed and efficiently bagged to save space.
Specifications

Specifications

General Purpose Lab Coat
Polypropylene
Snaps
Single-use
Knit
Triple over locked
Individually Packaged
Unisex
White
Knit
Chemicals
Yes
CE, EN 1513, EN 1149-1
Videos
SDS
Documents

Documents

Certificates