missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klercide™ 70/30 Denatured Ethanol
For use in effective disinfection procedure. Ecolab™ Klercide™ 70/30 Denatured Ethanol is sterilised by gamma irradiation.
POA - £937.00
Specifications
Product Type | Alcohol |
---|---|
Sterility | Sterile |
Filtered | Filtered |
For Use With (Application) | Effective disinfection procedure |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15356068
|
Ecolab™
3078260 |
500 mL |
£937.00
Pack of 12 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15386068
|
Ecolab™
3078300 |
1 L |
POA
Pack of 6 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Available as either 70/30 Isopropyl Alcohol (IPA) or Denatured Ethanol blended with water for injection (WFI) or deionized water
- Also available as 60% Isopropyl alcohol or Propan-1-ol and Denatured Ethanol, both blended with WFI
- Use pharma grade alcohol in blend
- Cleanroom processed, 0.2 micron filtered and terminally sterilized by gamma irradiation
- Available as spray systems fitted with SteriShield Delivery System, which ensures sterility of contents is preserved throughout use, aerosols, stream aerosol or in 5L capped container
- Available with complete supporting documentation
Specifications
Alcohol | |
Sterile | |
Filtered | |
Effective disinfection procedure |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Klercide 70/30 Denatured Ethanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : ethanol, 2-methylpropan-2-ol