Learn More
Ecolab Klercide 70/30 Denatured Ethanol Aerosol
Product Code. 17546526
Click to view available options
Sterility:
Sterile
Product Code. 17546526
Brand: Ecolab 3078380
Klercide 70/30 Denatured Ethanol blended with Water for Injection (WFI) with endotoxin level guaranteed to be less than 0.25 EU/mL. The alcohol blend is 0.2 micron filtered then filled and triple bagged in a Grade C cleanroom before being terminally sterilized by gamma irradiation.
  • Triple bagged for easy transfer into Grade A and B cleanrooms
  • Is gamma irradiated to ensure a sterile, spore free product
  • Is endotoxin tested to an approved pharmacopoeial method
Specifications
Product Type Disinfectant
Sterility Sterile
Composition Denatured Ethanol Blended with WFI