missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klercide™ 70/30 Denatured Ethanol Blended with WFI
For use in effective disinfection procedure. Ecolab™ Klercide™ 70/30 Denatured Ethanol Blended with WFI is sterilised by gamma irradiation.
Brand: Ecolab™ 3078360
181.20 GBP valid until 2024-12-20
Use promo code "21644" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available as either 70/30 Isopropyl Alcohol (IPA) or Denatured Ethanol blended with water for injection (WFI) or deionized water
- Also available as 60% Isopropyl alcohol or Propan-1-ol and Denatured Ethanol, both blended with WFI
- Use pharma grade alcohol in blend
- Cleanroom processed, 0.2 micron filtered and terminally sterilized by gamma irradiation
- Available as spray systems fitted with SteriShield Delivery System, which ensures sterility of contents is preserved throughout use, aerosols, a stream aerosol or in 5L capped container
- Available with complete supporting documentation
Specifications
Alcohol | |
Filtered | |
Disinfection procedure | |
1 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Klercide 70/30 Denatured Ethanol
Signal Word
- Danger||
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : ethanol, 2-methylpropan-2-ol