Learn More
Ecolab™ Klercide 70/30 Denatured Ethanol Blended with WFI
Keep your cleanroom free from contamination with this Klercide sterile alcohol. Ecolab™ Klercide 70/30 Denatured Ethanol Blended with WFI uses pharma grade alcohol in the blend. Spray systems fitted with the SteriShield Delivery System, preserving the sterility of the contents throughout use.
Brand: Ecolab™ 3078370
Code : NEW
Additional Details : CAS Number : 7732-18-5 Weight : 6.00000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- Manufactured in a state-of-the-art facility to the principles of cGMP
- Cleanroom processed, 0.2 micron filtered and terminally sterilized by gamma irradiation
- 70% denatured ethanol blended with Water for Injection (WFI)
- Endotoxin level guaranteed to be less than 0.25EU/mL
- Supplied sterile by gamma irradiation and triple bagged
Specifications
Denatured Ethanol with WFI | |
Sterile | |
70/30 Denatured Ethanol Blended with WFI | |
500 mL |
Safety and Handling
- Klercide 70/30 Denatured Ethanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : ethanol, 2-methylpropan-2-ol