Learn More
Ecolab Klercide 70/30 IPA Blended with Deionized Water
Klercide 70/30 IPA blended with deionized water is sterile and 0.2 micron filtered.
Brand: Ecolab 6600946
Additional Details : CAS Number : 67-63-0 Weight : 5.26500kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
- BPR approved
- Economical, high grade disinfection with easy application
- 0.2 micron filtered, filled and triple bagged in a Grade C (ISO Class 7) cleanroom
- Irradiated at no less than 25 kGy to ensure a sterile alcohol
- Supplied in a trigger spray incorporating the Sterishield Delivery System which ensures in-use sterility for up to 3 months
- The Sterishield Delivery System is the only fully validated closed system designed to keep the fluid contents of the bottle sterile at all times. This ensures full compliance with GMP Annex 1 requirements that disinfectant solutions are sterile prior to use
- Certificates of Analysis are available for every batch
- Full supporting technical documentation is available
Specifications
Disinfectant | |
Neutral | |
6 x 1 L |
Liquid | |
Sterile |
Safety and Handling
- Klercide 70/30 IPA
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol