Learn More
Ecolab™ Klercide™ Low Residue Quat with WFI
A very low residue quaternary ammonium compound and Water for Injection (WFI)
Brand:  Ecolab™ 3078730
Product Code. 15338488
  • £1165.00 / Pack of 4
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The biocide blend is 0.2 micron filtered and sterile filled under Grade A laminar flow and triple bagged in a Grade B cleanroom
  • A sterile ready-to-use daily disinfectant with low residue accumulation effect