Learn More
Ecolab™ Klercide Sporicidal Active Chlorine
Deisgned to control all types of microbial contamination in cleanrooms. Ecolab™ Klercide Sporicidal Active Chloring is an effective biocide based on active chlorine, available in a ready to use spray bottle.
Brand:  Ecolab™ 3078930
Product Code. 15352428
  • £633.00 / Pack of 6
Explore more special offers
Description
Description
  • Sterile by gamma irradiation
  • EN 1276 Bactericidal; EN 1650 Fungicidal (yeast); EN 13697 Bactericidal and fungicidal
  • Cleanroom processed
  • Unique Sterishield Delivery System ensures the sterility of the contents is preserved throughout use.
  • Available with complete supporting documentation
Specifications
Specifications
Biocide
Colorless
Spray Bottle
Bactericidal, fungicidal, sporicidal, virucidal agent
Sterile
Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and fungicidal EN 12397; Sporicidal EN 13704; Virucidal EN 14476
Klercide sporicidal active chlorine; Sporicidal Active Chlorine
1 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Klercide Sporicidal Active Chlorine
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : sodium hypochlorite