Learn More
Ecolab™ Klercide™ Sporicidal Low Residue Peroxide
A very low residue blend of high grade 6% hydrogen peroxide
Brand:  Ecolab™ 3079070
Product Code. 15336108
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • The biocide blend is 0.2 micron filtered and sterile filled under Grade A laminar flow and double bagged in a Grade B cleanroom
  • The product is a sterile ready-to-use rotational sporicide with very low residues for small and large surfaces
Specifications
Specifications
Peroxide Capped
5 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Klercide Sporicidal Low Residue Peroxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains :
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ecolab™ Klercide™ Sporicidal Low Residue Peroxide > Peroxide Capped