Learn More
Shield Medicare™ Klerwipe™ Sterile Dry Wipes
Contains no chemical binders so they do not leave undesirable residues. Shield Medicare™ Klerwipe™ Sterile Dry Wipes are made from a 68gsm, binder free, hydroentangled polyester/cellulose blend.
Brand:  Shield Medicare™ 3035510
Additional Details : Weight : 1.00000kg
Product Code. 12868040
  • £208.00 / Pack of 100
Estimated Shipment: 18-09-2024
to see stock.
Add to basket
Description
Description
Manufactured from 115gsm circular knit polyester with laser sealed edges. Low particulate wipes are made from a 68gsm binder free, hydroentangled cellulose/polyester blend.
  • Packed and double bagged in a Grade C (ISO Class 7) cleanroom before gamma irradiation at no less than 25kGy.
  • Suitable for use in controlled environments such as isolators and laminar flow cabinets.
  • All packaging is designed to minimise the generation of particles.
  • Suitable for all cleaning and disinfection procedures. The amount of cleaning agent can be varied to suit the particular purpose.
  • Cost effective
  • Packed in small quantities to avoid the need to compromise the sterility of a pack once opened.
  • Available in a wide range of sizes which eliminates wastage of unused wipes.
  • Highly absorbent Klerwipes are useful both for applying cleaning solutions and disinfectants and in cleaning up spills.
  • Suitable for direct product contact use.
Specifications
Specifications
Sterile Wiper
EN ISO 9001:2008, EN ISO 13485:2003 and European Medical Device Directive 93/42/EEC accredited company. Certificates of irradiation with every batch.
300 x 300 mm
Gamma-irradiated
Polyester
Hydroentangled polyester/cellulose blend
10 x 10 Pk.