Learn More
Klever Klever Excel Wide Cut
Recessed blade of the Klever Excel Wide Cut reduces cut injuries and damaged goods.
Brand:  Klever KCJ-4-30Y
 View more versions of this product
Product Code. 17728997
  • £230.00 / Pack of 50
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cuts boxes, banding, film, tape, and thousands more applications.
  • Multiple position handle; two positions for control and reach.
  • Available in both narrow and wide heads with durable, high quality carbon steel blade.
  • Innovative plastic tape splitter.
  • Advanced Plastic Polymers.
  • Highest Safety Level.
  • Disposable.
Specifications
Specifications
Klever Excel Wide Cutter
Single-use
Non-sterile
Yellow