missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14449-1-AP-150UL
This item is not returnable.
View return policy
Description
KLHL2 (kelch-like protein 2), also known as actin-binding protein Mayven, is a 593 amino acid actin-binding protein. Highly expressed in brain, KLHL2 is thought to play a role in the organization of the actin cytoskeleton in brain cells.Specifications
KLHL2 | |
Polyclonal | |
Unconjugated | |
KLHL2 | |
ABP KELCH, Actin binding protein Mayven, Kelch like protein 2, KLHL2, MAV, MAYVEN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11275, 290692, 77113 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
F1LZF0, O95198, Q8JZP3 | |
Klhl2 | |
KLHL2 Fusion Protein Ag5950 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |