missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Klotho-derived peptide 1
TGF-β receptor 2 (TβR2) binding peptide; disrupts the TGF-β/TβR2 interaction
Brand: Tocris Bioscience™ 7830/1
Additional Details : Weight : 0.00970kg
Description
Klotho-derived peptide 1 (KP1) is a peptide that binds to TGF-β receptor 2 (TβR2) (Kd = 1.4 μM). It inhibits TGF-β signaling by blocking TGF-β/TβR2 interaction. In mouse models of renal fibrosis, intravenous injection of KP1 results in its specific accumulation in the injured kidneys. It suppresses TGF-β signaling, repressing fibroblast activation and ameliorates kidney fibrosis in vivo.Specifications
9365 | |
Store at -20°C | |
M.W. (Observed): 3228.48 |
95% | |
EC 3.2.1, EC 3.2.1.31, KL, klotho, TGFB2, TGFbeta, TGFbeta 2 | |
1 mg |