Learn More
Spectrum Labs™ KMPi TFF System
Spectrum Labs™ KMPi TFF System is a tangential flow filtration system with process controls and data collection software for pilot-scale bioprocessing.
Brand:  Spectrum Labs™ SYM3-U20
Product Code. 15797586
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Ideal system for precise and reproducible control and documentation of tangential flow filtration processes ranging from 10 to 1000 Liters
  • Fully integrated system
    • Digital peristaltic pump
    • Multi-language interface with a graphical LCD display
    • Digital pressure monitor and automated alarm and shut-off controls
    • KMP Easy-Load pump head
    • Automatic backpressure valve, module stand, and real-time data collection and process control software
  • With the pump drive and Easy-Load pump Head, the KMP provides consistent and accurate flow rates with a range of 0.01 to 13.0 LPM
  • Two 13 LPM KMP Easy-Load pump heads or one 17 LPM high-performance pump head may also be used with the system
Specifications
Specifications
Tangential Flow Filtration
18 Kg
Complete Tangential Flow Filtration System
40 lbs.
Digitally controlled peristaltic pump, digital pressure monitor and transducers, automatic backpressure valve, KFComm data collection software, auxilliary scale, warranty (2 years)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Spectrum Labs™ KMPi TFF System > Complete tangential flow filtration system