missing translation for 'onlineSavingsMsg'
Learn More

KN-62, Tocris Bioscience™

Product Code. 30001223
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-competitive P2X7 antagonist

Selective, cell-permeable inhibitor of CaM kinase II (IC50 = 0.9 μM). Binds directly to the calmodulin binding site of the enzyme. Potent non-competitive antagonist at the P2X7 receptor (IC50 = 15 nM).

Specifications

CAS 127191-97-3
Molecular Formula C38H35N5O6S2
MDL Number MFCD00083180
Quantity 1 mg
Synonym s-4-2-n-methylisoquinoline-5-sulfonamido-3-oxo-3-4-phenylpiperazin-1-yl propyl phenyl isoquinoline-5-sulfonate, unii-63hm46xpow, chembl28324, 63hm46xpow, 1-n,o-bis 5-isoquinolinesulfonyl-n-methyl-l-tyrosyl-4-phenylpiperazine, s-5-isoquinolinesulfonic acid 4-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl ester, 4-2s-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl isoquinolinesulfonic acid ester, 4-2s-2-n-methylisoquinoline-5-sulfonamido-3-oxo-3-4-phenylpiperazin-1-yl propyl phenyl isoquinoline-5-sulfonate, 5-isoquinolinesulfonic acid, 4-2s-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl ester
InChI Key RJVLFQBBRSMWHX-UHFFFAOYNA-N
SMILES CN(C(CC1=CC=C(OS(=O)(=O)C2=C3C=CN=CC3=CC=C2)C=C1)C(=O)N1CCN(CC1)C1=CC=CC=C1)S(=O)(=O)C1=C2C=CN=CC2=CC=C1
IUPAC Name 4-[2-(N-methylisoquinoline-5-sulfonamido)-3-oxo-3-(4-phenylpiperazin-1-yl)propyl]phenyl isoquinoline-5-sulfonate
Molecular Weight (g/mol) 721.85
PubChem CID 5312126
Formula Weight 721.84
Percent Purity >98%
Chemical Name or Material KN-62
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.